missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids

Fit standard spectrophotometer cell holders

Manufacturer:  Fisherbrand™ 14958103

 View more versions of this product

Catalog No. 14-958-103




Description

Description

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Reduce reagent use with micro cells
  • Wavelength range for all cells is 200 to 2500nm
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

PTFE
Standard spectrophotometer cell holder
Ea., 2 Ea./Cs.
200 to 2500nm
12.5mm
Quartz
Micro Fluorescence Cell
For transmission from far ultraviolet through the infrared
45mm
Rectangular
2mm
100µL
10 mm
Documents
Promotions

Promotions