Please login to your online account to display your discounted pricing
Fisherbrand™ Flowmeter Regulators [GenStar]
For Carbon Dioxide, Nitrogen and Oxygen
Manufacturer: genstar technologies co  10575102
 View more versions of this product
Catalog No. 10-575-102
  • / Each
Description & Specifications
Specifications
Temperature Rating 0° to 140°C
Airflow Rate 5-32 SCFH
Internal Part Material Neoprene Fabric Diaphragm
For Use With (Application) Oxygen
Pressure Rating 80psig
Connections 0.62 in.-18RH(F) and 0.56 in.-18RH(M)
Seal Material Nylon
Case Material Brass