Please login to your online account to display your discounted pricing

Fisherbrand™ Fine Tip Marking Pens


Quick-drying ink offered in a variety of colors

Manufacturer: fisher scientific  140016FSC

 View more versions of this product

Catalog No. 13-379-5

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Product Type Securline™ Lab Marker
Quantity 10 Pack
Color Red
Latex Free Yes
For Use With (Application) Used to make permanent markings on plastic, metal, or porcelain labware
Sterile No
Tip Style Fine tip