Fisherbrand™ Fine Tip Marking Pens
Quick-drying ink offered in a variety of colors
Manufacturer:  Fisher Scientific 140020FSC
 View more versions of this product
Catalog No. 13-379-4
Specifications
Description
Pen, Marking; Fisherbrand; Fine tip; Fast drying; Ink is permanent on plastic, metal, and ceramic; Ink color: black; 10/Pk.
Designed to write on damp surfaces as easily as they write on dry surfaces. Proven permanent on most surfaces, including plastic, metal, and ceramic.
  • Fine tip is excellent for general marking
  • Highly resistant to abrasion and water
  • Will not smear when subjected to water or alcohol during normal use
Description & Specifications
Specifications
Used to make permanent markings on plastic, metal, or porcelain labware
Securline™ Lab Marker
No
Black
Yes
10 Pack
Fine tip