Fisherbrand™ Filters for TCLP
Binder-free, borosilicate glass fiber filters have 0.7 micron pore size
Manufacturer:  Fisherbrand™ SA1J078H00
 View more versions of this product
Catalog No. 09-804-142G
Specifications Description & Specifications
Specifications
5.59 in.
14.2cm
Borosilicate Glass Fiber
0.7μm
Ideal for samples with >10% solids, TCLP