Fisherbrand™ Filter Adapters
Eliminates boring of special size holes in rubber stoppers
Description
Eliminates boring of special size holes in rubber stoppers
Designed to fit Buchner and fritted-glass funnels, porcelain filters without headpieces, and reverse flow cleaning of filter crucibles. Useful in filtration assemblies for supporting funnels, Gooch crucibles, and filter tubes.
- Neoprene rubber
- Autoclavable
- Adapter does not need to be fitted closely around the funnel stem to be functional
- Can be used singly or nesting with adjacent sizes
- Available as an adapter set, or in individual sizes
Specifications
Autoclavable | |
Neoprene Rubber | |
0.15 in. | |
Filter Adapter | |
3.5 cm |
Size 5 | |
Buchner and fritted Glass funnels, porcelain filters without headpieces | |
0.4 cm | |
1.3 in. | |
58 x 38 mm |