Fisherbrand™ Filter Adapters
Eliminates boring of special size holes in rubber stoppers
Manufacturer:  Fisherbrand™ 1010A4
 View more versions of this product
Catalog No. 05-888-103
Description
Description
Eliminates boring of special size holes in rubber stoppers
Designed to fit Buchner and fritted-glass funnels, porcelain filters without headpieces, and reverse flow cleaning of filter crucibles. Useful in filtration assemblies for supporting funnels, Gooch crucibles, and filter tubes.
  • Neoprene rubber
  • Autoclavable
  • Adapter does not need to be fitted closely around the funnel stem to be functional
  • Can be used singly or nesting with adjacent sizes
  • Available as an adapter set, or in individual sizes
Specifications
Specifications
Autoclavable
46 x 29 mm
1.1 in.
0.1 in.
Filter Adapter
Size 4
Buchner and fritted Glass funnels, porcelain filters without headpieces
2.9 cm
0.35 cm
Neoprene Rubber