Fisherbrand™ Electroporation Cuvettes Plus™
Disposable, color-coded cuvets fit most electroporators. Sterilized by gamma irradiation and supplied individually wrapped.
Supplier: Fisherbrand™ 450151
Description
Disposable, color-coded cuvets are compatible with most electroporators.
- Clear, medical grade polycarbonate for high visibility
- Polished aluminum lowers arcing frequency
- Round cap allows single-handed removal
- FB102 has imbedded aluminum electrodes
Specifications
Sterile | |
400 μL | |
Imbedded aluminum electrodes | |
Electroporator |
Electroporation Cuvet | |
Polycarbonate | |
2 mm |