Fisherbrand™ Electroporation Cuvettes Plus™ Promo
Disposable, color-coded cuvets fit most electroporators
$360.48 - $360.48
Specifications
Material Polycarbonate
For Use With (Equipment) Electroporators
Sterility Sterile
Type Electroporation Cuvette
View More Specs
Products 1
Catalog Number Mfr. No. Width (Metric) Gap Capacity (Metric) Includes Price Quantity  
Catalog Number Mfr. No. Width (Metric) Gap Capacity (Metric) Includes Price Quantity  
98-549-8PM
View Documents
Fisherbrand™
985498
4mm 800 μL Imbedded aluminum electrodes; Pack of 50
Each for $360.48
Description
Description
  • Clear, medical grade polycarbonate for high visibility
  • Polished aluminum lowers arcing frequency
  • Round cap allows single-handed removal
  • Sterilized by gamma irradiation and supplied individually wrapped
  • 985491PM has embedded aluminum electrodes
Specifications
Specifications
Polycarbonate
Electroporators
Sterile
Electroporation Cuvette