Description
- Borosilicate glass, permanent amber markings, Tuf-Temp™-treated to reduce chipping
- Accuracy printed on each pipet
- Capacity marked below mouthpiece
Order Info
Available as PersonaPet™ personalized pipets
Compliance
Calibrated “To Deliver/Blow Out” (upper graduation) and “To Contain” (lower graduation).
Specifications
±0.003 mL | |
0.003 oz. | |
Dual Purpose Pipet |
Borosilicate Glass | |
0.1 mL |