missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Dry Block/Incubator Thermometers

Designed for use in all standard heating blocks and water baths

Manufacturer:  Hb Instrument B608101520

 View more versions of this product

Catalog No. 13201558


 

Specifications Description & Specifications

Specifications

±1°C
35mm
Arrows at 37, 56°C
225mm
1
Red Organic
Standard heating blocks and water baths
Partial
8.85 in.
White Back Glass, PFA coated
0° to 100°C
Accessories