Please login to your online account to display your discounted pricing
Fisherbrand™ Double Cytology Funnel with White Filter Cards
For the Thermo Scientific™ Shandon™ CytoSpin™ Centrifuge
Manufacturer: fisherbrand™  10-356*
 View more versions of this product
Catalog No. 10-356
  • / Pack of 25 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Includes White Filter Card; Sealing Caps
Quantity 10/Cs.
Volume (Metric) Up to 0.25mL
For Use With Shandon CytoSpin Centrifuge