missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Double Cytology Funnel with White Filter Cards
For the Shandon™ CytoSpin™ Centrifuge
$256.76 - $3,692.24
Specifications
For Use With (Equipment) | Shandon CytoSpin Centrifuge |
---|---|
Includes | White Filter Card; Sealing Caps |
Volume (Metric) | Up to 0.25 mL |
Description
- Functional equivalent of the Shandon Double Cytofunnel™
- Fully compatible with Shandon CytoSpin Centrifuge
- White filter card is pre-attached and is designed for single use only
- Funnel sealing caps included
- For sample volumes of up to 0.25mL or less
- Provides two deposition areas on each slide
Specifications
Shandon CytoSpin Centrifuge | |
White Filter Card; Sealing Caps | |
Up to 0.25 mL |