Fisherbrand™ Disposable Soda-Lime Glass Pasteur Pipets
Constriction near top prevents plugs from slipping into pipette body
Manufacturer:  Fisherbrand™ K63A136786A
 View more versions of this product
Catalog No. 13-678-6A
Description
Description
Constriction near top prevents plugs from slipping into pipette body
  • Uniform bore
  • Tip openings range from 0.80 to 1.20mm
  • Top and tip free of sharp edges
  • Can be cotton plugged (plugs not included)
  • Carton top folds back — tilts carton into convenient dispensing position
Specifications
Specifications
Yes
Uniform bore, free or sharp edge (tip and top)
14.6cm
2mL
For transferring samples to sample cups and for drop counting
5.75 in.
Pasteur Pipet
Soda Lime Glass
Documents
Accessories