missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Disposable Plastic Cuvets

Choice of polystyrene or methacrylate

$132.33 - $132.33



Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
14-386-20 Accessories Fisherbrand™
202295
Case of 500 Each for $132.33
Specifications

Description

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Select polystyrene cuvets for assays in visible spectral range (340 to 750nm)
  • For measurements in the visible and near-UV range (300-750nm) choose cuvets made from polymethyl methacrylate (acrylic)
  • Choice of 4.5 and 1.5mL sample capacity
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity

Order Info

  • Packaged 100 cuvets per stackable tray; five trays to a case
  • Outside dimensions: 12L × 12W × 45mmH