Fisherbrand™ Disposable Plain-End Glass Tubes with Color-Coded Label Area
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
 View more versions of this product
Catalog No. 14-957-26E
Description
Description
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
  • Fused-in, textured, color patches accept any type of ink or pencil
  • Expose to heat, steam, hours in a water bath, without affecting label
  • Marking area is large enough for all necessary information—1.37 in. (for 10 and 12mm O.D. tubes) or 2 in. (for 13 and 16mm O.D. tubes) long labeling area in five pastel colors
  • Round bottom
Specifications
Specifications
Blue
100 mm
Yes
Non-sterile
KIM-KAP
15 mL
Borosilicate Glass
End problem of lost or peeling labels
16 mm
Disposable Culture Tube