Please login to your online account to display your discounted pricing
Fisherbrand™ Disposable Pestle System
Low-cost alternative to mechanical or metal and glass tissue grinders
$30.10 - $30.10
Products
Catalog Number Mfr. No. Price Quantity    
03-392-107
fisherbrand™
F0002
Each for $30.10
Description & Specifications
Lightweight, cordless mixer can be used with disposable pestles and microtubes
  • High-speed vortexing action completely homogenizes cells and tissue within seconds; re-suspends protein and DNA pellets
  • Battery-powered unit greatly reduces hand fatigue
  • Grooved handle makes it easy to turn, even when wearing gloves
  • Rugged polybutylene terephthalate (PBTP) plastic pestle yields superior homogenization
  • Disposable pestles and tubes can be used by themselves or in conjunction with the grinder
  • Pestles and tubes are individually wrapped to prevent cross-contamination of samples
  • DNase, RNase, and pyrogen-free