Fisherbrand™ Disposable Heavy-Wall Borosilicate Glass Tubes
Chemically clean, fully annealed
Manufacturer:  Fisherbrand™ K60A1495810A
 View more versions of this product
Catalog No. 14-958-10A
Description
Description
Chemically clean, fully annealed
Order Info
Case of 1000 contains four lint-free trays
Specifications
Specifications
10mm
None
4mL
75mm
Disposable Culture Tube
Non-sterile
Yes
Tray
KIM-KAP
Borosilicate Glass
Round Bottom