Fisherbrand™ Disposable Flint Glass Tubes with Plain End
Specially annealed to reduce breakage along stress point
Manufacturer:  Fisherbrand™ K60G14958H
 View more versions of this product
Catalog No. 14-958H
Description
Description
Specially annealed to reduce breakage along stress point
  • Round bottom
  • Smooth top and bottom to prevent cuts
Specifications
Specifications
16mm
Specially annealed to reduce breakage along stress point
Tray
23mL
150mm
Disposable Culture Tube
Non-sterile
Yes
None
250/Tray
KIM-KAP
Flint Glass
Round Bottom