Thermo Scientific™ Disposable Filter Columns
Disposable polypropylene columns effectively filter out interfering materials from blood serum or urine
Manufacturer:  Thermo Scientific™ 1138750
Catalog No. 11-387-50
Description
Description
  • Save time and money Help eliminate inaccurate readings
  • To use, just hold screening column over sample cup
  • Decant directly through the column into sample cups — no pipetting
  • Filters 200 to 300μm When filtration is complete, discard column Sample is ready for analysis Columns are graduated in 0.5mL divisions
  • Serum filter column for final filtration directly into sample cups for Technicon™ 6/60™, 12/60™, and SMAC analyzers
  • Large flow channel and less capillarity let all the sample through Filters particles as small as 40 to 50μm Helps reduce line clogging
Specifications
Specifications
Polypropylene
200 to 300 μm
Yes
Filter Column
1000 Case
To filter out interfering materials from blood, serum, urine
Documents