Please login to your online account to display your discounted pricing
Fisherbrand Disposable Faceshields
High-quality, optically clear, distortion-free 7mil film.
Manufacturer: fisher science education  4101FEM
 View more versions of this product
Catalog No. S98126
  • / Pack of 25 Each
  • / Case of 4 Pack
Description & Specifications
Specifications
Certifications/Compliance OSHA 29 CFR 1910.1030, CDC
Film Thickness 7mil
Includes Stretch strap, Latex-free Foam band
Disposable Yes
Color Clear
Style Mid face
Material Polyester
Lens Material Polyester with anti-fog and Anti-static coating