Fisherbrand™ Disposable Cotton-Plugged Borosilicate-Glass Pasteur Pipets
Made of N-51A borosilicate glass.
Description
Made of N-51A borosilicate glass
- Straight-cut tips and smooth, fire-polished tops to facilitate bulb attachment
- A positive constriction at approximately 1 in. (2.5cm) from top holds supplied cotton plug in place, thereby reducing the chance of aspirating liquid into rubber bulb
- Easel box, made of nonlinting material, folds to become a dispenser for easy access to pipets
- Foam pad at bottom of box preserves tips
Specifications
Borosilicate Glass | |
5.75 in. | |
Smooth fire polished | |
14.6 cm | |
Single-use |
Nonsterile | |
Straight | |
2 mL | |
Cotton-plugged Pasteur Pipet |