Fisherbrand™ Disposable Auto-Analyzer Cups
Specifically designed for use with most clinical instrumentation
Manufacturer:  Fisherbrand™ 02544134
 View more versions of this product
Catalog No. 02-544-134
Specifications Description & Specifications
Specifications
0.5mL
Yes
Polystyrene
Round or Conical
Auto Analyzer Cup
Clear
Yes
Kodak Ektachem E400 and E700, Centrifichem and Technicon analyzer
Promotions
Promotions
  • Buy one case of Fisherbrand™ Auto Analyzer Cups and get one case at no additional cost. This is an NLSU exclusive offer. You must apply and qualify for the program to be eligible.
    06/30/18