Please login to your online account to display your discounted pricing

Fisherbrand™ Disposable Auto-Analyzer Cups


Specifically designed for use with most clinical instrumentation

Manufacturer: fisherbrand™  02544131

 View more versions of this product

Catalog No. 02-544-131

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Capacity (Metric) 2mL
Shape Round or Conical
For Use With (Equipment) Technicon, Tosoh AIA 600 and 1200 automatic analyzers, Kodak Ektachem E400 and E700 analyzer
Graduated Yes
Product Type Auto Analyzer Cup
Color Clear
Material Polystyrene
Disposable Yes