missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Dispensing Trays

Dispensing valve at one corner, close to bottom.

Manufacturer:  Fisherbrand™ 1088848

 View more versions of this product

Catalog No. 15-236-3H



Description

Description

Dispensing valve at one corner, close to bottom.

  • Durable 0.25 in. thick (6.4mm) high-density polyethylene tray
  • Welded corners and bottom

Order Info

Additional dispensing tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

White
8 in.
0.63cm
HDPE
20.25cm
40.6cm
16 x 16 x 8 in. (40.6 x 40.6 x 20.25cm)
16 in.
16 in.
Dispensing Tray
40.6cm
Documents