missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Dispenser Tips for Handystep™ S Repeating Pipetter

Manufactured from high-quality virgin plastics for use with all standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

Manufacturer:  Fisherbrand™ 702994

 View more versions of this product

Catalog No. 13-668-714



Description

Description

Manufactured from high-quality virgin plastics for use with all standard repeating pipettes in a wide variety of applications. The tips are made without chemical additives to prevent reagent contamination.

25mL and 50mL tips require the use of an adapter; One reusable adapter supplied in each package of 25mL and 50mL tips.

 

  • Precision seal between the piston and cylinder ensure smooth operation, increased accuracy and precision
  • Extended piston design minimizes dead volume and maximizes fluid utilization
  • Made entirely from high–quality virgin high–density polyethylene and polypropylene (Piston of 0.1mL tip is liquid crystal polymer-LCP)
Available sterile or nonsterile
  • Sterile: 13-668-711, -714, -715, -713, -716, -717, -718, -719, -721
  • Nonsterile: 13-668-700, -701, -702, -703, -704, -705, -708, -707, -709, -720 
Specifications

Specifications

HandyStep™ S, HandyStep™ Electronic, Eppendorf™ Repeater™, Rainin™ AutoRep™ E, Rainin™ AutoRep™ M, Gilson™ Repeater Pipettor
2.5mL
Non-Filtered
Translucent
≤0.8 to 0.2%
HDPE/PP (Polypropylene)
Individually Wrapped
50 to 250μL
Sterile
No
±≤3.5 to 0.7%
HandyStep™ S, HandyStep™ Electronic, Eppendorf™ Repeater™, Rainin™ AutoRep™ E, Rainin™ AutoRep™ M, Gilson™ Repeater Pipettor
Documents
Promotions

Promotions