Fisherbrand™ DISPENSE Filter System
Saves time, reduces lab costs and increases safety.
$120.85 - $203.15
Specifications
Product Type | Dispense Filtration System |
---|---|
For Use With (Application) | Dispensing samples for instrumentation, pour-off or other use |
For Use With (Equipment) | Use with Glass collection tubes |
Includes | Combination filter, transfer pipet, cap |
Quantity | 100 Pack |
Description
The restricted opening provides effective specimen closure and can help minimize exposure to droplets of blood or other potentially infectious materials during sample handling, preparation, processing, and storage.
- All-inclusive “closed” filtration system with a combination filter, transfer pipet, and cap
- Filters fibrin and gel particulates, dispenses serum or plasma, prevents evaporation, and helps protect ISE systems
- All-in-one system maintains sample integrity and eliminates transfer error
- Dispense minute or large amounts of sample without transfer pipets or add-on products
- Ideal for dispensing samples for instrumentation, pour-off or other use
- Use with glass collection tubes
Specifications
Dispense Filtration System | |
Use with Glass collection tubes | |
100 Pack |
Dispensing samples for instrumentation, pour-off or other use | |
Combination filter, transfer pipet, cap |