Please login to your online account to display your discounted pricing
Fisherbrand™ Diaphragm Pressure and Vacuum Gauges
Accuracy is ±1% in midrange, ±2 to 3% at extremes
Manufacturer: fisher scientific  G20840
 View more versions of this product
Catalog No. 11-281A
  • / Each
Description & Specifications
Specifications
Material Phosphor bronze
Product Type Pressure Gauge
Accuracy ±1% midrange, ±2 to 3% extreme