Please login to your online account to display your discounted pricing

Fisherbrand™ Delicate Dissecting Scissors


Forged from surgical-grade stainless steel

Manufacturer: fisherbrand™  SDI13015

 View more versions of this product

Catalog No. 08-951-5

  • / Each
 

Description & Specifications

Specifications

Blade Style Straight
Material Stainless Steel
Tip Style Straight
Grade Surgical
Autoclavable Yes
Length (English) 4.5 in.
Length (Metric) 11.43cm
Type Dissecting, Surgical, precision Scissors
For Use With (Application) Surgical, Precision