missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Data-Coded Hollow Cathode 2in. dia. Lamps for Perkin-Elmer™ Instruments

For Perkin-Elmer™ instruments

$1,004.30 - $1,004.30

Specifications

Diameter (English) Outer 2 in.
Diameter (Metric) Outer 5cm
Gas Fill Neon
Light Source Hollow Cathode
Product Type Spectrophotometer Lamp
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Amperage Material Wavelength Element Price Quantity  
Catalog Number Mfr. No. Amperage Material Wavelength Element Price Quantity  
14-386-119D Fisherbrand™
5UNCUP
20mA UV Glass 324.8nm Copper Each for $1,004.30
Specifications Description & Specifications

Specifications

2 in.
Neon
Spectrophotometer Lamp
500 hr.
5cm
Hollow Cathode
Perkin-Elmer™ instruments
T16 Shape