Please login to your online account to display your discounted pricing
Fisherbrand™ Custom Cleaned Clear Boston Rounds
Containers and caps are noncertified and precleaned with USP purified water
$128.53 - $128.53
Specifications
Cleaning Procedure USP purified Water
Closure Included Yes
Closure Liner Polyethylene
Closure Material PTFE
Closure Type screw cap
View More Specs
Products
Catalog Number Mfr. No. Capacity Closure Size Price Quantity    
02-912-011
fisherbrand™
02912011
4.22 oz., 125mL 22-400 Case of 24 Each for $128.53
Description & Specifications
Specifications
Cleaning Procedure USP purified Water
Closure Included Yes
Closure Liner Polyethylene
Closure Material PTFE
Closure Type screw cap
Color Clear
Material Glass
Shape Round
Type Boston round bottle
  • With attached PTFE-faced, PE-lined caps
  • Containers and caps are noncertified and precleaned with USP Purified Water for visible particulates in a clean process area
Custom packaging and certification available upon request