Fisherbrand™ Zahn Cup-Type Viscometers
Fisherbrand Zahn Cup-Type Viscometers are used for quick, accurate measurements of viscosity of liquids such as paint, lacquer, varnish, syrup, oil, etc.
Manufacturer:  Fisherbrand™ SDI2854850B
 View more versions of this product
Catalog No. 15-348-50B
Description
Description
  • Fisherbrand™ Zahn Cup-Type Viscometers are portable cups that measure the viscosity of a liquid in Zahn seconds (the amount of time required for a steady-stream to flow from the orifice at the bottom of the cup)
  • Measure viscosity of very thin to very heavy liquids
  • Ideal for unknown liquid experiments
  • Quick, accurate measurement
  • Made from ASTM™ Standard Stainless Steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation/autoclave
  • Calibration certificate, conversion charts and instructions included
Specifications
Specifications
1.5 oz.
1.5 oz., 44mL
0.27cm
35cm
For quick measurement of the viscosity of thin oil or lacquer
Corrosion Resistant Stainless Steel
44mL
0.108 in.
13.75 in.
40 to 230 cSt
Cup Type Viscometer No. 2
ASTM™
Documents