missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Cryo/Freezer Boxes

Choice of boxes and dividers to accommodate tubes from 0.5 to 50mL

Manufacturer:  Fisherbrand™ PSI398300

 View more versions of this product

Catalog No. 03-395-463


 

Requires : Boxes without dividers require your choice of Cat. Nos. 03-395-450, -452, -453, -457 to -462

Specifications

Description

Fiberbox, Cryo; Fisherbrand; With dividers; White; 3 x 3 x 2 in.; With 25 cells

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

2 in.
3 in.
3 in.
3 x 3 x 2 in. (7.6 x 7.6 x 5.1cm)
Cryo/Freezer Box
Used in mechanical freezers and liquid nitrogen tanks and all standard stainless steel and polycarbonate storage systems
25 cell dividers, bottom drain holes, tight lid
-196°C
5.1cm
7.6cm
7.6cm
White
Provides low-cost storage for laboratories in universities, government, medical research, and pharmaceutical facilities; in institutions performing artificial insemination, biotech, blood bank, and stem cell/bone marrow procedures
Fiberboard
-320.8°F