Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Manufacturer:  Fisherbrand™ ER45X502
 View more versions of this product
Catalog No. 12-543F
Specifications Description & Specifications
Specifications
Borosilicate Glass
50mm
0.17 to 0.25mm
Coverglass
Rectangular
45mm
Microscope, histology, cytology, hematology, microbiology