Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Manufacturer:  Fisherbrand™ ER22X601.5
 View more versions of this product
Catalog No. 12-544G
Specifications Description & Specifications
Specifications
Borosilicate Glass
60mm
0.16 to 0.19mm
Coverglass
Rectangular
22mm
Microscope, histology, cytology, hematology, microbiology