Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Manufacturer:  Fisherbrand™ ER22X301.5
 View more versions of this product
Catalog No. 12-544A
Specifications Description & Specifications
Specifications
Coverglass
22mm
30mm
Microscope, histology, cytology, hematology, microbiology
0.16 to 0.19mm
Borosilicate Glass
Rectangular