Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Manufacturer:  Fisherbrand™ ER22X601
 View more versions of this product
Catalog No. 12-545J
Specifications Description & Specifications
Specifications
Borosilicate Glass
60mm
0.13 to 0.17mm
Coverglass
Rectangular
22mm
Microscope, histology, cytology, hematology, microbiology