Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Manufacturer:  Fisherbrand™ ER24X601
 View more versions of this product
Catalog No. 12-545M
Specifications Description & Specifications
Specifications
Coverglass
60mm
Rectangular
Borosilicate Glass
24mm
0.13 to 0.17mm
Microscope, histology, cytology, hematology, microbiology