Fisherbrand™ Cover Glasses: Rectangles
Made of finest optical borosilicate glass, with uniform thickness and size
Manufacturer:  Fisherbrand™ ER35X501
 View more versions of this product
Catalog No. 12-545G
Specifications Description & Specifications
Specifications
Borosilicate Glass
50mm
0.13 to 0.17mm
Coverglass
Rectangular
35mm
Microscope, histology, cytology, hematology, microbiology