Fisherbrand™ Cork Supports
Durable, compressed cork lab rings
Manufacturer:  Fisherbrand™ 55004
 View more versions of this product
Catalog No. 07-835C
Description
Description
Durable, compressed cork lab rings
  • Concave recesses support dishes and round-bottom flasks
  • Durable
Specifications
Specifications
12 cm
4.72 in.
17 cm
1.18 in.
Cork
Not Autoclavable
6.69 in.
Concave recesses support dishes and 5000mL round bottom flasks
3 cm