missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Cork Stoppers - Laboratory Grade

100% natural cork

Manufacturer:  Fisherbrand™ 50335

 View more versions of this product

Catalog No. 07-782F



Description

Description

100% natural cork

  • Punched out from natural cork wood
  • Tapered to size
  • Safe to use in the packaging and/or processing of food

Specifications

Specifications

Ideal for sealing bottles, safe to use in the packaging and/or processing of food
500/Pk.
Natural
0.56 in.
1.9cm
Stopper
Laboratory
3
1.06cm
1.42cm
Wood
3
SDS
Documents
Product Certifications
Accessories