missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Cork Stoppers - Laboratory Grade
100% natural cork
Manufacturer: Fisherbrand™ 54134
Description
100% natural cork
- Punched out from natural cork wood
- Tapered to size
- Safe to use in the packaging and/or processing of food
Specifications
Ideal for sealing bottles, safe to use in the packaging and/or processing of food | |
100/Pk. | |
Natural | |
0.78cm | |
Wood |
Laboratory | |
00 | |
0.31 in. | |
1.27cm |