missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Cork Stoppers - Laboratory Grade
100% natural cork
$9.69 - $163.28
Specifications
Color | Natural |
---|---|
Material | Wood |
Product Type Specs | Stopper |
For Use With (Application) | Ideal for sealing bottles, safe to use in the packaging and/or processing of food |
Grade | Laboratory |
Catalog Number | Mfr. No. | Stopper Number | Length (Metric) | Diameter (Metric) Bottom | Quantity | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Stopper Number | Length (Metric) | Diameter (Metric) Bottom | Quantity | Price | Quantity | ||
07-782A
|
Fisherbrand™
53935 |
36586 | 1.27cm | 0.4cm | 500/Pk. |
Pack of 500 Each for $46.07
|
|
07-782B
|
Fisherbrand™
54135 |
0 | 1.27cm | 0.55cm | 500/Pk. |
Pack of 500 Each for $46.07
|
|
07-782D
|
Fisherbrand™
50135 |
1 | 1.58cm | 0.83cm | 500/Pk. |
Pack of 500 Each for $50.07
|
|
07-782E
|
Fisherbrand™
50235 |
2 | 1.74cm | 0.96cm | 500/Pk. |
Pack of 500 Each for $54.66
|
|
07-782F
|
Fisherbrand™
50335 |
3 | 1.9cm | 1.06cm | 500/Pk. |
Pack of 500 Each for $58.93
|
|
07-782G
|
Fisherbrand™
50435 |
4 | 2.06cm | 1.19cm | 500/Pk. |
Pack of 500 Each for $60.90
|
|
07-782H
|
Fisherbrand™
50535 |
5 | 2.22cm | 1.34cm | 500/Pk. |
Pack of 500 Each for $62.52
|
|
07-782J
|
Fisherbrand™
50635 |
6 | 2.38cm | 1.47cm | 500/Pk. |
Pack of 500 Each for $65.62
|
|
07-782K
|
Fisherbrand™
50735 |
7 | 2.54cm | 1.6cm | 500/Pk. |
Pack of 500 Each for $72.17
|
|
07-781A
|
Fisherbrand™
53934 |
36586 | 1.27cm | 0.4cm | 100/Pk. |
Pack of 100 Each for $9.69
|
|
07-781B
|
Fisherbrand™
54134 |
0 | 1.27cm | 0.55cm | 100/Pk. |
Pack of 100 Each for $9.69
|
|
07-781C
|
Fisherbrand™
50034 |
0 | 1.27cm | 0.71cm | 100/Pk. |
Pack of 100 Each for $9.69
|
|
07-781D
|
Fisherbrand™
50134 |
1 | 1.58cm | 0.83cm | 100/Pk. |
Pack of 100 Each for $11.00
|
|
07-781E
|
Fisherbrand™
50234 |
2 | 1.74cm | 0.96cm | 100/Pk. |
Pack of 100 Each for $11.55
|
|
07-781F
|
Fisherbrand™
50334 |
3 | 1.9cm | 1.06cm | 100/Pk. |
Pack of 100 Each for $11.03
|
|
07-781G
|
Fisherbrand™
50434 |
4 | 2.06cm | 1.19cm | 100/Pk. |
Pack of 100 Each for $12.52
|
|
07-781H
|
Fisherbrand™
50534 |
5 | 2.22cm | 1.34cm | 100/Pk. |
Pack of 100 Each for $13.03
|
|
07-781J
|
Fisherbrand™
50634 |
6 | 2.38cm | 1.47cm | 100/Pk. |
Pack of 100 Each for $13.41
|
|
07-781K
|
Fisherbrand™
50734 |
7 | 2.54cm | 1.6cm | 100/Pk. |
Pack of 100 Each for $14.62
|
|
07-781L
|
Fisherbrand™
50834 |
8 | 2.69cm | 1.7cm | 100/Pk. |
Pack of 100 Each for $19.86
|
|
07-781M
|
Fisherbrand™
50934 |
9 | 2.85cm | 1.85cm | 100/Pk. |
Pack of 100 Each for $23.79
|
|
07-781N
|
Fisherbrand™
51034 |
10 | 3.17cm | 1.95cm | 100/Pk. |
Pack of 100 Each for $38.45
|
|
07-781P
|
Fisherbrand™
51134 |
11 | 3.17cm | 2.1cm | 100/Pk. |
Pack of 100 Each for $45.41
|
|
07-781Q
|
Fisherbrand™
51234 |
13 | 3.17cm | 2.26cm | 100/Pk. |
Pack of 100 Each for $57.31
|
|
07-781R
|
Fisherbrand™
51334 |
14 | 3.17cm | 2.41cm | 100/Pk. |
Pack of 100 Each for $63.45
|
|
07-781S
|
Fisherbrand™
51434 |
14 | 3.17cm | 0.43cm | 100/Pk. |
Pack of 100 Each for $72.90
|
|
07-781T
|
Fisherbrand™
51534 |
15 | 3.17cm | 1.07 in. | 100/Pk. |
Pack of 100 Each for $78.72
|
|
07-781U
|
Fisherbrand™
51634 |
16 | 3.81cm | 2.76cm | 100/Pk. |
Pack of 100 Each for $97.66
|
|
07-781V
|
Fisherbrand™
51734 |
17 | 3.81cm | 2.92cm | 100/Pk. |
Pack of 100 Each for $101.62
|
|
07-781W
|
Fisherbrand™
51834 |
18 | 3.81cm | 3.07cm | 100/Pk. |
Pack of 100 Each for $122.76
|
|
07-781Y
|
Fisherbrand™
52034 |
20 | 3.81cm | 3.4cm | 100/Pk. |
Pack of 100 Each for $163.28
|
|
07-782C
|
Fisherbrand™
50035 |
0 | 1.27cm | 0.71cm | 500/Pk. |
Pack of 500 Each for $46.07
|
|
Description
100% natural cork
- Punched out from natural cork wood
- Tapered to size
- Safe to use in the packaging and/or processing of food
Specifications
Natural | |
Stopper | |
Laboratory |
Wood | |
Ideal for sealing bottles, safe to use in the packaging and/or processing of food |