missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Cork Stoppers - Laboratory Grade

100% natural cork

Manufacturer:  Fisherbrand™ 51534

 View more versions of this product

Catalog No. 07-781T



Description

Description

100% natural cork

  • Punched out from natural cork wood
  • Tapered to size
  • Safe to use in the packaging and/or processing of food

Specifications

Specifications

Ideal for sealing bottles, safe to use in the packaging and/or processing of food
100/Pk.
Natural
3.33cm
Wood
Laboratory
15
3.33cm
3.17cm
SDS
Documents