missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Cork Stoppers - Laboratory Grade

100% natural cork

Manufacturer:  Fisherbrand™ 51434

 View more versions of this product

Catalog No. 07-781S



Description

Description

100% natural cork

  • Punched out from natural cork wood
  • Tapered to size
  • Safe to use in the packaging and/or processing of food

Specifications

Specifications

Ideal for sealing bottles, safe to use in the packaging and/or processing of food
100/Pk.
Natural
1.25 in.
3.17cm
Stopper
Laboratory
14
0.43cm
3.17cm
Wood
14
SDS
Documents
Product Certifications