missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Laboratory Grade Cork Stoppers, Assorted Sizes
$48.03 - $48.03
Specifications
Grade | Laboratory |
---|---|
Material | Wood |
For Use With (Application) | Ideal for sealing bottles |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
07-785
|
Fisherbrand™
55122 |
6 to 15 |
Pack of 100 Each for $48.03
|
|
Description
- Choice of two grades: laboratory grade (corks cut from selected wood), or superior grade (corks cut from high-quality wood)
- Assorted lengths; Sizes: 6-15
Specifications
Laboratory | |
Wood | |
Ideal for sealing bottles |