Fisherbrand™ ColorFrost™ Plus Microscope Slides
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
 View more versions of this product
Catalog No. 12-550-16A
Description
Description
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
Same outstanding features as regularColorFrost slides with special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings.
  • Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
  • Colored marking surface for easy identification throughout procedures
  • Size: 25 × 75mm
Specifications
Specifications
Microscope Slide
Yes
25 mm
Glass
75 mm
Tan
SDS
Documents
Documents
Accessories