Please login to your online account to display your discounted pricing

Fisherbrand™ Colored Screw Caps


For use with nonsterile skirted- and conical-bottom tubes

Manufacturer: fisherbrand™  525RFIS

 View more versions of this product

Catalog No. 02-681-361

  • / Pack of 500 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Autoclavable Yes
Material Polypropylene
Color Red
Product Type Screw Cap with O-ring
For Use With (Equipment) Non-sterile skirted and conical bottom tubes
Includes O-ring