Please login to your online account to display your discounted pricing

Fisherbrand™ Colored Screw Caps

For use with nonsterile skirted- and conical-bottom tubes

Manufacturer: fisherbrand™  524-V-FIS

 View all options for this product

Catalog No. 02-681-354

 

Description & Specifications

Specifications

Autoclavable Yes
Material Polypropylene
Color Violet
Product Type Screw Cap without O-ring
Sterile Yes
For Use With (Equipment) Non-sterile skirted and conical bottom tubes

  • Feature a frosted writing surface
  • Can be pierced for sample removal