Please login to your online account to display your discounted pricing

Fisherbrand™ Colored Screw Caps


For use with nonsterile skirted- and conical-bottom tubes

Manufacturer: fisherbrand™  525AFIS

 View more versions of this product

Catalog No. 02-681-367

  • / Pack of 500 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

For Use With (Equipment) Non-sterile skirted and conical bottom tubes
Material Polypropylene
Product Type Screw Cap with O-ring
Autoclavable Yes
Includes O-ring
Color Amber